Assay Details
| Gene Symbol: RAB7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078013.1 | NP_001071481.1 | 1 - 2 | 183 | 100 | |
| GenBank mRNA | BC123789.1 | - | 1 - 2 | 183 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123789.1, NM_001078013.1 transcripts.
Set Membership: |
|

RAB7B
Genomic Map