Assay Details
| Gene Symbol: RAB7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078013.1 | NP_001071481.1 | 4 - 5 | 600 | 153 | |
| GenBank mRNA | BC123789.1 | - | 4 - 5 | 600 | 153 | |
More Information
Set Membership: |
|

RAB7B
Genomic Map