Assay Details
| Gene Symbol: MYOZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078039.1 | NP_001071507.1 | 2 - 3 | 297 | 100 | |
| GenBank mRNA | BC123727.1 | - | 2 - 3 | 297 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123727.1, NM_001078039.1 transcripts.
Set Membership: |
|

MYOZ3
Genomic Map