Assay Details
| Gene Symbol: MYOZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078039.1 | NP_001071507.1 | 3 - 4 | 455 | 76 | |
| GenBank mRNA | BC123727.1 | - | 3 - 4 | 455 | 76 | |
More Information
Set Membership: |
|

MYOZ3
Genomic Map