Assay Details
| Gene Symbol: PMEPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078078.1 | NP_001071546.1 | 3 - 4 | 288 | 68 | |
| GenBank mRNA | BC123607.1 | - | 3 - 4 | 288 | 68 | |
More Information
Set Membership: |
|

PMEPA1
Genomic Map