Assay Details
| Gene Symbol: MPIG6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078081.2 | NP_001071549.1 | 1 - 2 | 81 | 121 | |
| GenBank mRNA | BC120063.1 | - | 1 - 2 | 83 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120063.1, NM_001078081.2 transcripts.
Set Membership: |
|

MPIG6B
Genomic Map