Assay Details
| Gene Symbol: MPIG6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078081.2 | NP_001071549.1 | 5 - 6 | 642 | 75 | |
| GenBank mRNA | BC120063.1 | - | 5 - 6 | 644 | 75 | |
More Information
Set Membership: |
|

MPIG6B
Genomic Map