Assay Details
| Gene Symbol: PDRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078115.1 | NP_001071583.1 | 3 - 4 | 281 | 95 | |
| GenBank mRNA | BC118162.1 | - | 3 - 4 | 281 | 95 | |
More Information
Set Membership: |
|

PDRG1
Genomic Map