Assay Details
| Gene Symbol: NRBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078131.1 | NP_001071599.1 | 3 - 4 | 324 | 66 | |
| GenBank mRNA | BC123769.1 | - | 3 - 4 | 324 | 66 | |
More Information
Set Membership: |
|

NRBF2
Genomic Map