Assay Details
| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079511.2 | NP_001072979.1 | 1 - 2 | 153 | 124 | |
| GenBank mRNA | BC122777.1 | - | 1 - 2 | 154 | 124 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122777.1, NM_001079511.2 transcripts.
Set Membership: |
|

MCRIP2
Genomic Map