Assay Details
| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079511.2 | NP_001072979.1 | 4 - 5 | 493 | 123 | |
| GenBank mRNA | BC122777.1 | - | 4 - 5 | 494 | 123 | |
More Information
Set Membership: |
|

MCRIP2
Genomic Map