Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079579.1 | NP_001073047.1 | 2 - 3 | 237 | 82 | |
| GenBank mRNA | BC122678.1 | - | 2 - 3 | 237 | 82 | |
More Information
Set Membership: |
|

GRAMD2B
Genomic Map