Assay Details
| Gene Symbol: TMEM120A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079600.1 | NP_001073068.1 | 1 - 2 | 124 | 62 | |
| GenBank mRNA | BC122849.1 | - | 1 - 2 | 124 | 62 | |
More Information
Set Membership: |
|

TMEM120A
Genomic Map