Assay Details
| Gene Symbol: WDR83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079650.2 | NP_001073118.1 | 5 - 6 | 563 | 101 | |
| GenBank mRNA | BC120088.1 | - | 5 - 6 | 564 | 101 | |
More Information
Set Membership: |
|

WDR83
Genomic Map