Assay Details
| Gene Symbol: HAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079781.2 | NP_001073249.1 | 1 - 2 | 167 | 64 | |
| GenBank mRNA | BC120064.1 | - | 1 - 2 | 170 | 64 | |
More Information
Set Membership: |
|

HAO1
Genomic Map