Assay Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079795.1 | NP_001073263.1 | 3 - 4 | 533 | 131 | |
| GenBank mRNA | BC118330.1 | - | 3 - 4 | 575 | 131 | |
| BT026226.1 | - | 3 - 4 | 533 | 131 | ||
More Information
Set Membership: |
|

TMEM267
Genomic Map