Assay Details
| Gene Symbol: ARHGEF39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079800.1 | NP_001073268.1 | 2 - 3 | 285 | 64 | |
| GenBank mRNA | BC120169.1 | - | 2 - 3 | 285 | 64 | |
| BT026317.1 | - | 2 - 3 | 236 | 64 | ||
More Information
Set Membership: |
|

ARHGEF39
Genomic Map