Assay Details
| Gene Symbol: TRMT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080273.2 | NP_001073742.1 | 11 - 12 | 1151 | 91 | |
| GenBank mRNA | BC122733.1 | - | 11 - 12 | 1153 | 91 | |
More Information
Set Membership: |
|

TRMT11
Genomic Map