Assay Details
| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080309.2 | NP_001073778.1 | 1 - 2 | 356 | 73 | |
| GenBank mRNA | BC122788.1 | - | 1 - 2 | 357 | 73 | |
More Information
Set Membership: |
|

YOD1
Genomic Map