Assay Details
| Gene Symbol: LXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080340.2 | NP_001073809.1 | 3 - 4 | 722 | 96 | |
| GenBank mRNA | BC120149.1 | - | 3 - 4 | 723 | 96 | |
More Information
Set Membership: |
|

LXN
Genomic Map