Assay Details
| Gene Symbol: TAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080371.2 | NP_001073840.2 | 5 - 6 | 666 | 62 | |
| GenBank mRNA | BT026536.1 | - | 4 - 5 | 461 | 62 | |
More Information
Set Membership: |
|

TAB3
Genomic Map