Assay Details
| Gene Symbol: KRT28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080375.1 | NP_001073844.1 | 7 - 8 | 1245 | 119 | |
| GenBank mRNA | BC118316.1 | - | 7 - 8 | 1245 | 119 | |
More Information
Set Membership: |
|

KRT28
Genomic Map