Assay Details
| Gene Symbol: FSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081518.1 | NP_001074987.1 | 9 - 10 | 973 | 56 | |
| GenBank mRNA | BC122630.1 | - | 9 - 10 | 973 | 56 | |
More Information
Set Membership: |
|

FSD1
Genomic Map