Assay Details
| Gene Symbol: LEO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082604.1 | NP_001076073.1 | 8 - 9 | 1482 | 64 | |
| GenBank mRNA | BC120399.1 | - | 8 - 9 | 1482 | 64 | |
More Information
Set Membership: |
|

LEO1
Genomic Map