Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173939.2 | NP_776364.2 | 11 - 12 | 2135 | 97 | |
| GenBank mRNA | AJ300476.1 | - | 10 - 11 | 1950 | 97 | |
| BC122629.1 | - | 11 - 12 | 2135 | 97 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map