Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173939.2 | NP_776364.2 | 5 - 6 | 1266 | 80 | |
| GenBank mRNA | AJ300476.1 | - | 4 - 5 | 1081 | 80 | |
| BC122629.1 | - | 5 - 6 | 1266 | 80 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map