Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173939.2 | NP_776364.2 | 7 - 8 | 1627 | 103 | |
| GenBank mRNA | AJ300476.1 | - | 6 - 7 | 1442 | 103 | |
| BC122629.1 | - | 7 - 8 | 1627 | 103 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map