Assay Details
| Gene Symbol: RAB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174447.4 | NP_776872.1 | 1 - 2 | 85 | 55 | |
| GenBank mRNA | BC122723.1 | - | 1 - 2 | 86 | 55 | |
| M19886.1 | - | 1 - 2 | 41 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122723.1, NM_174447.4 transcripts.
Set Membership: |
|

RAB3B
Genomic Map