Assay Details
| Gene Symbol: RAB3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174447.4 | NP_776872.1 | 2 - 3 | 318 | 68 | |
| GenBank mRNA | BC122723.1 | - | 2 - 3 | 319 | 68 | |
| M19886.1 | - | 2 - 3 | 274 | 68 | ||
More Information
Set Membership: |
|

RAB3B
Genomic Map