Assay Details
| Gene Symbol: DNAJB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174532.3 | NP_776957.2 | 3 - 4 | 307 | 80 | |
| GenBank mRNA | AF426743.1 | - | 2 - 3 | 221 | 80 | |
| BC122622.1 | - | 3 - 4 | 307 | 80 | ||
| BT021624.1 | - | 3 - 4 | 301 | 80 | ||
More Information
Set Membership: |
|

DNAJB6
Genomic Map