Assay Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174739.4 | NP_777164.1 | 2 - 3 | 117 | 51 | |
| GenBank mRNA | AF393210.1 | - | 3 - 4 | 345 | 51 | |
| BC120055.1 | - | 2 - 3 | 118 | 51 | ||
More Information
Set Membership: |
|

CBY1
Genomic Map