Assay Details
| Gene Symbol: VKORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003903.3 | NP_001003903.1 | 1 - 2 | 232 | 68 | |
| GenBank mRNA | AY682746.1 | - | 1 - 2 | 173 | 68 | |
| BC133442.1 | - | 1 - 2 | 234 | 68 | ||
More Information
Set Membership: |
|

VKORC1
Genomic Map