Assay Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010992.7 | NP_001010992.2 | 1 - 2 | 234 | 82 | |
| GenBank mRNA | AY858551.2 | - | 1 - 2 | 234 | 82 | |
More Information
Set Membership: |
|

DIO2
Genomic Map