Assay Details
| Gene Symbol: LTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013401.2 | NP_001013419.1 | 2 - 3 | 215 | 87 | |
| GenBank mRNA | BC114670.1 | - | 2 - 3 | 215 | 87 | |
More Information
Set Membership: |
|

LTA
Genomic Map