Assay Details
| Gene Symbol: URGCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015513.2 | NP_001015513.2 | 4 - 5 | 233 | 80 | |
| GenBank mRNA | BC126571.1 | - | 4 - 5 | 234 | 80 | |
| BT021657.1 | - | 4 - 5 | 221 | 80 | ||
More Information
Set Membership: |
|

URGCP
Genomic Map