Assay Details
| Gene Symbol: RASL11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015635.2 | NP_001015635.1 | 1 - 2 | 326 | 79 | |
| GenBank mRNA | BC142345.1 | - | 1 - 2 | 339 | 79 | |
| BT020927.1 | - | 1 - 2 | 258 | 79 | ||
| BT029844.1 | - | 1 - 2 | 326 | 79 | ||
More Information
Set Membership: |
|

RASL11B
Genomic Map