Assay Details
| Gene Symbol: PLSCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046053.2 | NP_001039518.1 | 1 - 2 | 443 | 106 | |
| GenBank mRNA | BC114676.1 | - | 1 - 2 | 91 | 106 | |
| BT029864.1 | - | 1 - 2 | 443 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114676.1, NM_001046053.2 transcripts.
Set Membership: |
|

PLSCR3
Genomic Map