Assay Details
| Gene Symbol: ACOT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075157.2 | NP_001068625.2 | 2 - 3 | 342 | 65 | |
| GenBank mRNA | BC114814.1 | - | 2 - 3 | 342 | 65 | |
More Information
Set Membership: |
|

ACOT8
Genomic Map