Assay Details
| Gene Symbol: SCGB1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076976.3 | NP_001070444.2 | 1 - 2 | 112 | 102 | |
| GenBank mRNA | AY994053.1 | - | 1 - 2 | 120 | 102 | |
| BC126820.1 | - | 1 - 2 | 113 | 102 | ||
More Information
Set Membership: |
|

SCGB1A1
Genomic Map