Assay Details
| Gene Symbol: BMF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077485.1 | NP_001070953.1 | 2 - 3 | 454 | 63 | |
| GenBank mRNA | AB238943.1 | - | 2 - 3 | 454 | 63 | |
More Information
Set Membership: |
|

BMF
Genomic Map