Assay Details
| Gene Symbol: NFE2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077899.2 | NP_001071367.2 | 2 - 3 | 965 | 102 | |
| GenBank mRNA | BC134642.1 | - | 2 - 3 | 965 | 102 | |
More Information
Set Membership: |
|

NFE2L3
Genomic Map