Assay Details
| Gene Symbol: CPA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077905.2 | NP_001071373.1 | 7 - 8 | 1009 | 57 | |
| GenBank mRNA | BC126678.1 | - | 7 - 8 | 1010 | 57 | |
More Information
Set Membership: |
|

CPA5
Genomic Map