Assay Details
| Gene Symbol: LYZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077910.1 | NP_001071378.1 | 1 - 2 | 59 | 66 | |
| GenBank mRNA | BC126794.1 | - | 1 - 2 | 59 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126794.1, NM_001077910.1 transcripts.
Set Membership: |
|

LYZL1
Genomic Map