Assay Details
| Gene Symbol: LYZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077910.1 | NP_001071378.1 | 2 - 3 | 227 | 71 | |
| GenBank mRNA | BC126794.1 | - | 2 - 3 | 227 | 71 | |
More Information
Set Membership: |
|

LYZL1
Genomic Map