Assay Details
| Gene Symbol: ERRFI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077930.1 | NP_001071398.1 | 1 - 2 | 155 | 78 | |
| GenBank mRNA | BC126579.1 | - | 1 - 2 | 155 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126579.1, NM_001077930.1 transcripts.
Set Membership: |
|

ERRFI1
Genomic Map