Assay Details
| Gene Symbol: PMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077981.2 | NP_001071449.1 | 2 - 3 | 236 | 69 | |
| GenBank mRNA | BC126839.1 | - | 2 - 3 | 237 | 69 | |
More Information
Set Membership: |
|

PMS1
Genomic Map