Assay Details
| Gene Symbol: SPART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077996.1 | NP_001071464.1 | 2 - 3 | 951 | 99 | |
| GenBank mRNA | BC126710.1 | - | 2 - 3 | 951 | 99 | |
More Information
Set Membership: |
|

SPART
Genomic Map