Assay Details
| Gene Symbol: SPART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077996.1 | NP_001071464.1 | 7 - 8 | 1777 | 82 | |
| GenBank mRNA | BC126710.1 | - | 7 - 8 | 1777 | 82 | |
More Information
Set Membership: |
|

SPART
Genomic Map