Assay Details
| Gene Symbol: ZBTB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078016.1 | NP_001071484.1 | 1 - 2 | 155 | 97 | |
| GenBank mRNA | BC126550.1 | - | 1 - 2 | 155 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126550.1, NM_001078016.1 transcripts.
Set Membership: |
|

ZBTB18
Genomic Map