Assay Details
| Gene Symbol: SLC7A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078019.2 | NP_001071487.1 | 6 - 7 | 1203 | 76 | |
| GenBank mRNA | BC126655.1 | - | 6 - 7 | 1204 | 76 | |
More Information
Set Membership: |
|

SLC7A3
Genomic Map